missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol

Catalog No. AC119690010
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Plastic bottle
Plastic Jerrycan
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119690010 1 L Plastic bottle
AC119690025 2.5 L Plastic bottle
AC119690050 5 L Plastic Jerrycan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC119690010 Supplier Thermo Scientific Chemicals Supplier No. 119690010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant. | CAS: 50-00-0 | CH2O | 30.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; that can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol

Applications

  • It May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant

Specifications

Chemical Name or Material Formaldehyde
Name Note 37 wt% Solution in Water, Stabilized with 10 to 15% Methanol
CAS 50-00-0, 7732-18-5, 67-56-1
Melting Point -15.0°C
Color Clear
Density 1.0830g/mL
Boiling Point 97.0°C
Flash Point 50°C
Packaging Plastic bottle
Molecular Formula CH2O
Refractive Index 1.3755 to 1.3775
Linear Formula HCHO
MDL Number MFCD00003274
Quantity 1 L
Fieser 01,397; 02,200; 04,238; 05,312; 06,264; 07,158; 08,231; 09,224; 10,186; 11,240; 12,232; 14,168; 15,161; 16,171
Merck Index 15, 4263
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and acetone
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES C=O
IUPAC Name formaldehyde
Stabilizer 5 to 15% methanol
Molecular Weight (g/mol) 30.03
PubChem CID 712
ChEBI CHEBI:16842
Vapor Pressure 2mbar at 20°C
Formula Weight 30.03
Percent Purity 35.0 to 39.0%
Physical Form Solution
Show More Show Less

RUO – Research Use Only