missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde 37%, StatLab™

Catalog No. STL285305
Change view
Click to view available options
Packaging:
Cube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
STL285305 Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STL285305 Supplier Statlab Supplier No. 285305*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, if in contact with skin, or if inhaled.

37% Formaldehyde formula used to prepare formalin solutions.

  • Contains methanol as a stabilizer

Specifications

CAS 50-00-0
Packaging Cube
Quantity 5 gal.
Concentration or Composition (by Analyte or Components) 37% Formaldehyde, 63% Methyl Alcohol