missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 10% (v/v), Buffered, pH 7.0, Histological Fixative, Ricca Chemical

Catalog No. 31908
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31908 250 mL
319032 1 L
31901 4 L
319025 10 L
31905 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31908 Supplier Ricca Chemical Company Supplier No. 31908
Only null left

Specifications

Packaging Type Amber Glass Bottle
Concentration 10% (v/v)
For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Formaldehyde
Quantity 250 mL
Solution Type Formaldehyde
Form Solution