missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 10% (v/v) (non-buffered), Histological Fixative, Ricca Chemical

Catalog No. 318032
Change view
Click to view available options
Capacity:
1 L
4 L
Packaging Type:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Packaging Type
318032 1 L Natural Poly Bottle
31801 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 318032 Supplier Ricca Chemical Company Supplier No. 318032
Only null left

Specifications

Capacity 1 L
Packaging Type Natural Poly Bottle
For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Formaldehyde
Form Solution