Learn More
StatLab FormaGuard Neutralizing Pad
Description
The FormaGuard™ Pad is light blue, rigid, polycoated, and roughly 1/8 in. in thickness
Specifications
Specifications
| Material | Pre-treated Formalin Neutralizer |
| Type | Formalin Neutralizer |
| Format | Pad |
| Dimensions (L x W) | 15 x 20 in. |