missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Forma™ Freezer Replacement Shelves
Description
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
- Stainless steel
- Kits include mounting hardware
Specifications
Specifications
| Type | Shelf Kit |
| Capacity (English) | 13 cu. ft. |
| Capacity (Metric) | 368.1 L |
| Style | Upright |
| Color | Silver |
| For Use With (Equipment) | 13.0 cu. ft. Freezers |