missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Form Module Sistema Elix

For use with Elix

Supplier:  MilliporeSigma™ ZLX0EDI10

Catalog No. ZLX0EDI10


Only null left
Explore available promotions

Description

Description

Water purification accessory
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications

Specifications

Form Module Sistema Elix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions