Learn More
Fork Resonance Apparatus
Description
Demonstrates acoustical resonance, audible beats, and sympathetic vibration. Two heavy, chrome-plated steel, 256Hz tuning forks, a rubber mallet, two resonance boxes (30 x 12 x 27cm), and two slider masses.
- Slider masses cause frequency of the tuning fork to decrease by approximately 8% when positioned on uppermost part of fork
- Changing position of the boxes and slider masses allow different frequencies and beats to be heard
Specifications
Specifications
| Product Type | Fork Resonance Apparatus |
| For Use With (Application) | Demonstrate acoustical resonance, audible beats and sympathetic vibration |
| Includes | Two heavy, Chrome plated Steel, a Rubber mallet, 2 slider masses |
| Frequency | 256 Hz |