Learn More
Global Life Sciences (Formerly Pall Lab) Forceps, Stainless Steel with Multi-Colored Grips (3/pk)
Supplier: Cytiva PALL4690
For easy placement of membrane filters into filter funnels or petri dishes.Flat surfaceAvoid membrane damageStainless steelTips have a flat, smooth surface to avoid membrane filter damage. Stainless steel construction with polypropylene finger grips for a comfortable and secure hold. Autoclavable. This product is not approved for use in medical, clinical, surgical or other patient applications.