Learn More
Sklar Instruments Econo(TM) Sterile Kelly Hemostatic Forceps, Sklar , 5-1/2 in., Blunt, Curved, Serrated, Stainless Steel, Disposable, Sterile. CS/50. DFS Item
Supplier:  Sklar Instruments 962562
Econo(TM) Sterile Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Blunt, Curved, Serrated, Stainless Steel, Disposable, Sterile. Econo(TM) Sterile Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Econo(TM) Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments. BX50.
Catalog No. NC9414448
Only null left
Explore available promotions