missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Food Red No. 40, TCI America™
Specifications
Specifications
| Color | Red/Yellow |
| Quantity | 25 g |
| Physical Form | Powder |
| Formula Weight | 496.42 |
| Chemical Name or Material | Food Red No. 40 |
Safety and Handling
| RTECSNumber | QK2260000 |
| TSCA | Yes |