missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Food Red No. 106, TCI America™
Specifications
Specifications
| Color | Purple |
| Quantity | 25 g |
| Physical Form | Powder |
| Formula Weight | 580.65 |
| Chemical Name or Material | Food Red No. 106 |
Safety and Handling
| EINECSNumber | (5)-1504 |
| RTECSNumber | BP6750000 |
| TSCA | Yes |