missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Food Red No. 102, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Physical Form | Powder |
| Formula Weight | 604.46 |
| Chemical Name or Material | Food Red No. 102 |
Safety and Handling
| EINECSNumber | (5)-1495 |
| RTECSNumber | QJ6530000 |
| TSCA | Yes |