missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Food Green No. 3, TCI America™
Specifications
Specifications
| Color | Green |
| Quantity | 10 g |
| Physical Form | Powder |
| Formula Weight | 808.84 |
| Chemical Name or Material | Food Green No. 3 |
Safety and Handling
| EINECSNumber | (5)-5228 |
| RTECSNumber | BQ4425000 |
| TSCA | Yes |