missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Food Calorimeter 

Allows students to determine the amount of energy released through the burning of food. EISCO Food Clirometer is an easy method involves the measuring of the change in temperature of a sample of water from the combustion of a small amount of food.

Supplier:  Eisco™ PH42715F

Catalog No. S23759

Only null left
Explore available promotions

Description

Description

Allows students to determine the amount of energy released through the burning of food. EISCO Food Clirometer is an easy method involves the measuring of the change in temperature of a sample of water from the combustion of a small amount of food.

  • Experiment introduces concepts in thermodynamics and leads to an understanding of energy and its relation to food consumption
  • Experiment Guide included with product

Kit contains:

  • Erlenmeyer Flask (125mL)
  • Cylindrical Metal Stand: Dia x H: 4 x 6.75 in.
  • Metal suspension plate 4.5 x 4.5 in.
  • Cork with nail (Food sample Holding assembly)
  • Rubber stopper

Specifications

Specifications

Food Calorimeter
Determine the amount of energy released through the burning of food.
  • Erlenmeyer Flask (125mL)
  • Cylindrical Metal Stand: Dia x H: 4 x 6.75 in.
  • Metal suspension plate 4.5 x 4.5 in.
  • Cork with nail (Food sample Holding assembly)
  • Rubber stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions