missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Food Blue No. 2, TCI America™
Supplier: TCI America F014825G
Specifications
| Food Blue No. 2 | |
| C16H8N2Na2O8S2 | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate | |
| 5284351 | |
| Blue/Purple | |
| Powder |
| 860-22-0 | |
| MFCD00005723 | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O | |
| 466.35 | |
| 466.35 | |
| 25 g |
Chemical Identifiers
| 860-22-0 | |
| 466.35 | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate |
| C16H8N2Na2O8S2 | |
| MFCD00005723 | |
| 5284351 | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
Safety and Handling
EINECSNumber : (5)-1650&(5)-1732
RTECSNumber : DU3000000
TSCA : Yes