missing translation for 'onlineSavingsMsg'
Learn More

Fomblin YR-1800, Thermo Scientific Chemicals

Catalog No. AAL1754806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1754806 5 g
AAL1754814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1754806 Supplier Thermo Scientific Chemicals Supplier No. L1754806

It is used in applications include pump and compressor cleaning, degreasing, dewatering, vapor blanketing, flash point reduction, and trace residue elimination. It acts as a diluent in the application of more viscous PFPE fluids and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69991-67-9
Molecular Formula C3F6
Molecular Weight (g/mol) 150.023
MDL Number MFCD01323324
InChI Key HCDGVLDPFQMKDK-UHFFFAOYSA-N
Synonym hexafluoropropylene, hexafluoropropene, perfluoropropene, perfluoropropylene, propene, hexafluoro, perfluoro-1-propene, propylene, hexafluoro, hexfluoropropylene, 1-propene, 1,1,2,3,3,3-hexafluoro, 1,1,2,3,3,3-hexafluoro-1-propene
PubChem CID 8302
IUPAC Name 1,1,2,3,3,3-hexafluoroprop-1-ene
SMILES C(=C(F)F)(C(F)(F)F)F

Specifications

Density 1.92
Solubility Information Difficult to mix in water.
Quantity 5 g
Refractive Index 1.304
Chemical Name or Material Fomblin YR-1800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only