Learn More
Folinic acid calcium salt hydrate, 98%
CAS: 1492-18-8 | C20H21CaN7O7 | 511.51 g/mol
$273.62 - $826.16
Chemical Identifiers
| CAS | 1492-18-8 |
|---|---|
| Molecular Formula | C20H21CaN7O7 |
| Molecular Weight (g/mol) | 511.51 |
| MDL Number | MFCD00006704 |
| InChI Key | KVUAALJSMIVURS-AEQORCIRNA-L |
| Synonym | calcium folinate |
| PubChem CID | 131675590 |
| IUPAC Name | calcium;(4S)-4-[[4-[(2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate |
| SMILES | [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH60181MD
|
Thermo Scientific Chemicals
H60181MD |
250 mg |
Each for $273.62
|
|
|||||
|
AAH6018103
|
Thermo Scientific Chemicals
H6018103 |
1 g |
Each for $826.16
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1492-18-8 | |
| 511.51 | |
| KVUAALJSMIVURS-AEQORCIRNA-L | |
| 131675590 | |
| [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1 |
| C20H21CaN7O7 | |
| MFCD00006704 | |
| calcium folinate | |
| calcium;(4S)-4-[[4-[(2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate |
Specifications
| 1492-18-8 | |
| MFCD00006704 | |
| 5723924 | |
| KVUAALJSMIVURS-AEQORCIRNA-L | |
| calcium;(4S)-4-[[4-[(2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate | |
| 131675590 | |
| 98% |
| C20H21CaN7O7 | |
| 250 mg | |
| calcium folinate | |
| [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1 | |
| 511.51 | |
| 511.51 (Anhydrous) | |
| Folinic acid calcium salt hydrate |
Safety and Handling
GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
EINECSNumber : 216-082-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only