missing translation for 'onlineSavingsMsg'
Learn More

Folin and Ciocalteu's Phenol Reagent, 2.0N, MP Biomedicals

Catalog No. icn19518690
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19518690 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19518690 Supplier MP Biomedicals Inc Supplier No. 0219518690
Only null left

Specifications

Chemical Name or Material Folin and Ciocalteu's Phenol Reagent
Name Note 2.0N
CAS 10102-25-7, 10102-40-6, 7647-01-0, 10213-10-2, 7664-38-2
Quantity 500 mL