missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folic Acid, Powder, USP, 97-102%, Spectrum™ Chemical
Description
Spectrum™ Chemical Folic Acid, Powder, USP is a water soluble B vitamin used to treat folic acid deficiency associated with ulcerative colitis, alcoholism, kidney dialysis, and liver disease and certain types of anemia. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.3% |
| Packaging | Amber Glass Bottle |
| Quantity | 100 g |
| Percent Purity | 97 to 102% |
| Grade | USP |