missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folic Acid Impurity D Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Folic Acid Impurity D is an impurity of the water-soluble B-vitamin, folic acid. Folic Acid is generally used in the treatment of certain types of anemia.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Specifications
Specifications
| CAS | 119-24-4 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C14H12N6O3 |
| MDL Number | MFCD00075823 |
| Synonym | Pteroic acid |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 312.28 |
| Grade | Certified Reference Material |