missing translation for 'onlineSavingsMsg'
Learn More

Folic acid, crystalline

Catalog No. p-7041956
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6083314 25 g
AAJ6083322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6083314 Supplier Thermo Scientific Chemicals Supplier No. J6083314
Only null left

CAS: 59-30-3 | C19H19N7O6 | 441.404 g/mol

Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid acts as a vitamin (hematopoietic), a nutritional supplement and dietary requirement for poultry. It also plays an important role in the formation of S-adenosyl methionine and in the synthesis and repair of DNA and RNA. Further, it serves as an essential vitamin for erythropoiesis. It is also employed in the treatment of methanol poisoning. In addition to this, it is used in the treatment and prevention of folate deficiencies and megaloblastic anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid acts as a vitamin (hematopoietic), a nutritional supplement and dietary requirement for poultry. It also plays an important role in the formation of S-adenosyl methionine and in the synthesis and repair of DNA and RNA. Further, it serves as an essential vitamin for erythropoiesis. It is also employed in the treatment of methanol poisoning. In addition to this, it is used in the treatment and prevention of folate deficiencies and megaloblastic anemia.

Solubility
Soluble in sodium hydroxide, acetic acid, phenol, pyridine, alkali hydroxides, carbonates, hot dilute hydrochloric acid and sulfuric acid. Slightly soluble in water, methanol, ethanol and butanol. Insoluble in acetone, chloroform, ether and benzene.

Notes
Light and heat sensitive. Incompatible with heavy metal ions, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.404
MDL Number MFCD00079305
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N

Specifications

Color Yellow to Orange
Quantity 25 g
Beilstein 100781
Merck Index 14,4221
Solubility Information Soluble in sodium hydroxide,acetic acid,phenol,pyridine,alkali hydroxides,carbonates,hot dilute hydrochloric acid and sulfuric acid. Slightly soluble in water,methanol,ethanol and butanol. Insoluble in acetone,chloroform,ether and benzene.
Formula Weight 441.41
Physical Form Crystalline
Chemical Name or Material Folic acid
EINECSNumber 200-419-0
RTECSNumber LP5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only