missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Folic acid, 97%, pure

Catalog No. p-3758052
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC216630500 50 g Glass bottle
AC216632500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC216630500 Supplier Thermo Scientific Chemicals Supplier No. 216630500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59-30-3 | C19H19N7O6 | 441.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.4
MDL Number MFCD00079305
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N

Specifications

Melting Point 250.0°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Merck Index 15, 4248
Specific Rotation Condition + 20.00 (20.00°C c=1,0.1N NaOH)
Specific Rotation + 20.00
Solubility Information Solubility in water: 1.6mg/l (25°C). Other solubilities: soluble to about 1% in boiling water,soluble in diluted acids and alkalis,soluble in hot diluted hcl and h2so4,relatively soluble in acetic acid,phenol,pyridin,of alkali hydroxides and ca
Formula Weight 441.4
Percent Purity 97%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Folic acid, Pure
Show More Show Less
EINECSNumber 200-419-

RUO – Research Use Only