missing translation for 'onlineSavingsMsg'
Learn More

Folic acid, ≥95%, Cell Culture Reagent Grade, MP Biomedicals™

$68.65 - $68.65

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.40
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19466505
View Documents
MP Biomedicals Inc
0219466505
5 g
Each for $68.65
Only null left
 
Description

Description

Specifications

Chemical Identifiers

59-30-3
441.40
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
CHEBI:27470
C19H19N7O6
OVBPIULPVIDEAO-LBPRGKRZSA-N
6037

Specifications

59-30-3
5 g
OVBPIULPVIDEAO-LBPRGKRZSA-N
6037
C19H19N7O6
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
441.40
CHEBI:27470
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'