missing translation for 'onlineSavingsMsg'
Learn More

Folic acid, 95-102%

Catalog No. p-7076599
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6293706 5 g
AAJ6293714 25 g
AAJ6293722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6293706 Supplier Thermo Scientific Chemicals Supplier No. J6293706
Only null left

CAS: 59-30-3 | C19H19N7O6 | 441.404 g/mol

Folic acid and its derivatives are essential mediators of one-carbon metabolism within cells.It is especially important in aiding rapid cell division and growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.404
MDL Number MFCD00079305
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N

Specifications

Packaging Glass bottle
Quantity 5 g
Beilstein 100781
Solubility Information Soluble in water (1.6mg/ml),1 M NaOH (50mg/ml),methanol (slightly),diluted acids,and alkaline solutions.
Formula Weight 441.41
Physical Form Powder
Chemical Name or Material Folic acid
EINECSNumber 200-419-0
RTECSNumber LP5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only