missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Folic Acid
Description
Folic Acid
Specifications
Specifications
| CAS | 471-53-4 |
| Melting Point | 292°C to 295°C |
| Molecular Formula | C30H46O4 |
| Linear Formula | C30H46O4 |
| MDL Number | MFCD00003706 |
| Quantity | 10 g |
| Synonym | PteGlu; Pteroyl-L-glutamic acid; Vitamin M |
| Molecular Weight (g/mol) | 470.68 g/mol |