missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Folic Acid
Description
Folic Acid
Spécifications
Spécifications
| CAS | 59-30-3 |
| Melting Point | > 285°C |
| Molecular Formula | C19H19N7O6 |
| Linear Formula | C19H19N7O6 |
| MDL Number | MFCD00079305 |
| Quantity | 25 g |
| Synonym | PteGlu; Pteroyl-L-glutamic acid; Vitamin M |
| Molecular Weight (g/mol) | 441.4 g/mol |