Learn More
Keystone Cleanroom Products Cleanroom Face Masks, Gamma Irradiated with Tyvek™ Side Extension
Sterile cleanroom facemasks feature an tyvek side extension for additional protection. Available with ties or double elastic headband fastening.
Supplier:  Keystone Cleanroom Products S1461XLGAD
 Afficher plus de versions de ce produit
Numéro de catalogue. 19722016
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Sterile cleanroom face masks
  • Certificates of Irradiation and Conformance with each shipment
  • Tyvek™ side extensions allow mask expansion to increase coverage and prevent unwanted facial skin exposure
Spécifications
Spécifications
Cleanroom Face Mask
Ties
X-Large
12.25 x 4 in.
Class 100 (ISO 5)
Side Extension
12.25 in.
S-1461
10 cm
Green
SMS
Sterile
Pleated
Single-use
Yes
31 cm
4 in.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS