Learn More
Foamtec UltraSOLV™ ScrubDISKs
Catalog No. 19304805
Encompass
Change view
Click to view available options
3000 Diamond Grit; Backed with pressure sensitive adhesive
Catalog No.
19304805 3000 Diamond Grit; Backed with pressure sensitive adhesive
1 options
Catalog No. 19304805 Supplier Foamtec Supplier No. HT4530PDC3G 1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Wide grit selection allows for quick removal of process-induced residue from aluminum, stainless steel, ceramic, glass, quartz and anodized surfaces and minimizes tool wear.
  • Patented scrubbing fiber free tool
  • Fiber-free construction for reducing particle levels in cleaned vacuum chambers
  • Unique bonding technology for minimizing release of abrasive particles
  • Eliminates H2O2 from PMs
  • Removes hardened process residue from Hi vacuum chambers
  • For use in conjunction with ErgoSCRUB™ hand tool