Learn More
Foamtec UltraSOLV™ ScrubDISKs DFS Item
Supplier:  Foamtec HT4520PDC3G1
Catalog No. 19-304-804
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Wide grit selection allows for quick removal of process-induced residue from aluminum, stainless steel, ceramic, glass, quartz and anodized surfaces and minimizes tool wear.
  • Patented scrubbing fiber free tool
  • Fiber-free construction for reducing particle levels in cleaned vacuum chambers
  • Unique bonding technology for minimizing release of abrasive particles
  • Eliminates H2O2 from PMs
  • Removes hardened process residue from Hi vacuum chambers
  • For use in conjunction with ErgoSCRUB™ hand tool
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Product Suggestions
Product Suggestions