Learn More
Foamtec UltraSOLV™ ScrubDISKs
Supplier: Foamtec HT4514DC3G1
Description
Wide grit selection allows for quick removal of process-induced residue from aluminum, stainless steel, ceramic, glass, quartz and anodized surfaces and minimizes tool wear.
- Patented scrubbing fiber free tool
- Fiber-free construction for reducing particle levels in cleaned vacuum chambers
- Unique bonding technology for minimizing release of abrasive particles
- Eliminates H2O2 from PMs
- Removes hardened process residue from Hi vacuum chambers
- For use in conjunction with ErgoSCRUB™ hand tool