Learn More
Foamtec UltraMOP™ Self-Wringing Mop Heads, Sterile DFS Item
Self-wringing sterile sponge mop head with stainless steel bracket
Supplier:  Foamtec FS600ST12
 View more versions of this product
Catalog No. 19-304-571
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
14 in.
Autoclavable
Designed for wet mopping procedures in sterile and controlled environments
Non-sterile
Stainless Steel Bracket
Polyurethane
Product Suggestions
Product Suggestions