Learn More
Foamtec PolyCLEAN™ Wipers DFS Item
Lightweight, knitted-wiper constructed and processed to meet clean room requirements
Supplier:  Foamtec HT5290L
Catalog No. 19-304-711
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Constructed from a continuous-filament fiber to resist shedding particles due to abrasion, making it the perfect choice for applications where cleanliness must be matched with cost control.
  • Washed and processed in a Class 10 environment making it ideal for particle sensitive wiping applications
  • Available in a black fabric for inspection and cleaning verification procedures
  • Amide and Silicone free for disk drive applications
  • Cut, washed and packed in our state-of-the-art integrated wiper facility to ensure product consistency
Specifications
Specifications
Polyclean™ cleanroom wipes
22.86 cm
22.86 cm
131
Black
9 x 9 in. (22.86 x 22.86 cm)
9 in.
9 in.
Polyester Fabric
ISO9001, ISO13485
Laser cut
For medical device and pharmaceutical clean and disinfections applications
Product Suggestions
Product Suggestions