Learn More
Foamtec PharmaMOP™ Cleanroom Mops
For critical cleanroom surfaces allowing the operator to clean ceilings, walls and floors with ease
Supplier:  Foamtec FT416D
Encompass
 View more versions of this product
Catalog No. 19304763
Description
Description
PharmaMOP utilizes simple press fit connection that enables gloved operators to easily attach mop heads and is compatible with steam sterilization cycles. Mop heads enable the quick, efficient cleaning of surfaces stained with disinfectant residues.
  • Two direction 360° swivel and mop head
  • Abrasion resistant cleanroom fabric
  • Class 100 processing
  • Fiber-free mopping of rough, non-skid floors
  • Sterile
  • Ultra-lightweight telescoping handle
Certifications
ISO 9001:2000 & 13485:2003 certified.
Specifications
Chemical Identifiers
ISO 9001:2000, ISO 13485:2003
Sterile
Critical cleanroom surfaces allowing the operator to clean ceilings, walls and floors with ease
Specifications
16 in.
Locking head frame, lever lock on telescoping handle
ISO 9001:2000, ISO 13485:2003
Sterile
Critical cleanroom surfaces allowing the operator to clean ceilings, walls and floors with ease
Product Suggestions
Product Suggestions