Learn More
Foamtec MiraWIPE™ Wipers DFS Item
Suitable for disinfection of vacuum chambers, medical devices and cleanrooms
Supplier:  Foamtec HT5711S25ST
 View more versions of this product
Catalog No. 19-304-562
Only null left
Description
Description
MiraWIPE™ wipers feature a non-symmetrical micro fiber structure allows them to scrub microbial biofilms and sporicidal residues.
  • Made of continuous filament micro denier, super soft polyester/nylon textile
  • High absorbency and contamination pickup
  • Abrasion resistant, no-run, no-stretch fabric to minimize In-Use Particles from the face of the wiper
  • Available in configurations to meet Class 1-1000 applications
Specifications
Specifications
Cleanroom Wipe
27.9 cm
27.9 cm
ISO Class 4
11 x 11 in. (27.9 x 27.9 cm)
25 Wipers/Bag
11 in.
11 in.
Polyester
Wide sealed
For removing stubbornly adhered particles and stains in pharmaceutical and micro electronics cleaning applications, ISO Class 10
Sterile
Product Suggestions
Product Suggestions