missing translation for 'onlineSavingsMsg'
Learn More

FMOC-D-phenylalanine, 98%

Catalog No. AC441710010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441710010 1 g
AC441710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441710010 Supplier Thermo Scientific Chemicals Supplier No. 441710010

CAS: 86123-10-6 | C24H21NO4 | 387.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86123-10-6
Molecular Formula C24H21NO4
Molecular Weight (g/mol) 387.44
MDL Number MFCD00062955
InChI Key SJVFAHZPLIXNDH-JOCHJYFZSA-N
Synonym fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid
PubChem CID 978332
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
SMILES OC(=O)[C@@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 184.0°C to 186.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation + 40.00
Formula Weight 387.43
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material FMOC-D-phenylalanine
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only