missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Fluotrac™ 600 High Binding Flat Bottomed Microplates

Description
- Non-pyrogenic
- Conical well design with flat bottom
- Ensures accurate pipetting
- Reduces dead volume of well
- Free of detectable DNase, RNase, human DNA
- Prevents wicking
Specifications
Specifications
| Barcode | Alphanumeric |
| Certified | Free of human DNA and pyrogen |
| Format | Standard |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Binding Type | High Binding |
| Color | Black |
| Product Type | Assay Microplate |
| Material | Polystyrene |
| Volume (Metric) Well | 25 to 340 μL |
| Show More |