missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One FLUOTRAC™ 200 96-well Non-Treated Microplate
GREENER_CHOICE

Catalog No. 07000722
Change view
Click to view available options
Color:
Black
White
Binding Property:
High Binding
Medium Binding
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Binding Property
07000722 Black High Binding
07000721 Black Medium Binding
07000085 White Medium Binding
07000083 White High Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07000722 Supplier Greiner Bio-One Supplier No. 675077

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in fluorescence immunoassay. Greiner Bio-One FLUOTRAC™ 200 96-Well Non-Treated Microplate is suitable for non polar proteins and peptides.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Can be traced back to production through defined LOT number system
  • Reduction of sample volume by up to 50 %

Service Plan

No

Warranty

No

Specifications

Barcode Alphanumeric
Binding Property High Binding
Format Half-Area
Array 8 x 12
Binding Type High Binding
Color Black
Product Type Assay Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Untreated
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Fluorescence immunoassay
No. per Case 40
Quantity 40/Cs.
RNase Free Yes
Show More Show Less