missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoSpheres™ Polystyrene Microspheres, 10 μm, blue fluorescent (365/415), for blood flow determination

Catalog No. f8829
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F8829 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F8829 Supplier Invitrogen™ Supplier No. F8829
Only null left

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene.

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Our Molecular Probes™ FluoSpheres™ beads are manufactured using high-quality, ultraclean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy.

FluoSpheres™ microspheres in the 10–15μm size range were designed specifically for regional blood flow studies in tissues and organs. Typically for these types of studies, the microspheres are injected at desired locations in the circulatory system and eventually lodge in the capillaries, where they can be extracted and the fluorescence quantified. The use of fluorescent microspheres eliminates the radiation hazards and disposal hurdles associated with radiolabeled microspheres. Multiple (up to seven or more with good spectral correction) simultaneously circulating FluoSpheres™ microspheres can be discriminated. Microspheres that have been perfused or injected into tissues and organs retain their fluorescence following histological serial sectioning. Our FluoSpheres™ microspheres for blood flow determination, which are available in eleven distinguishable fluorescent colors, are also compatible with blood flow analyzer systems that perform automated extraction and analysis.

FluoSpheres™ Microsphere Specifications:
• Label (Ex/Em): Blue fluorescent (365/415)
• Nominal bead diameter: 10 μm
• Coupling surface: Carboxylate

Characteristics of the Carboxylate Coupling Surface
Carboxylate-modified FluoSpheres™ beads have a high density of pendent carboxylic acids on their surface, making them suitable for covalent coupling of proteins and other amine-containing biomolecules using water-soluble carbodiimide reagents such as EDAC.

Choices for FluoSpheres™ Fluorescent Microspheres
In addition to microspheres for blood flow studies, investigate our complete offering of fluorescent microspheres products. Among those products you’ll find beads with these variations:
• Ten fluorescent colors
• Ten nominal bead diameters: 0.02 μm, 0.04 μm, 0.1 μm, 0.2 μm, 0.5 μm, 1.0 μm, 2.0 μm, 4.0 μm, 10.0 μm, and 15.0 μm
• Four surface modifications for protein coupling: carboxylate, sulfate, aldehyde-sulfate, amine
• Microspheres that are additionally precoupled with streptavidin, NeutrAvidin, biotin, europium, and platinum

Choices for Unstained Microspheres
We also offer hundreds of choices for UltraClean™ surfactant-free microspheres for research and commercial applications.

We’ll Make a Custom Microsphere Product for You
We will prepare custom orders upon request. For example, FluoSpheres™ beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Specifications

Concentration 3.6 x 106 beads/mL
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Color Blue
Material Polystyrene
Product Type Microsphere
Product Line FLUOSPHERES
Quantity 10 mL
Shipping Condition Room Temperature
Diameter (Metric) 10 μm

For Research Use Only. Not for use in diagnostic procedures.