missing translation for 'onlineSavingsMsg'
Learn More

Fluosilicic acid, 25 wt.% aqueous solution

CAS: 16961-83-4 | F6H2Si | 144.091 g/mol

$80.65 - $80.65

Specifications

Chemical Name or Material Fluosilicic acid
Color Colorless
CAS 16961-83-4,7732-18-5
Density 1.2700g/mL
Packaging Plastic bottle
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC410715000
View Documents
Thermo Scientific Chemicals
410715000
500 g Plastic bottle
Each for $80.65
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Fluosilicic acid
16961-83-4,7732-18-5
Plastic bottle
F6H2Si
Liquid
hexafluorosilicic acid, fluorosilicic acid, dihydrogen hexafluorosilicate, silicate 2-, hexafluoro-, dihydrogen, hexafluorosilicon 2-; hydron, dihydrogen hexafluorosilanediuide, silicofluoridd, hexafluorosilic acid, hydrofluorosilic acid, kieselfluorwasserstoffsaure
[H+].[H+].F[Si-2](F)(F)(F)(F)F
144.091
144.09
Colorless
1.2700g/mL
1.27
H2SiF6
MFCD00036289
OHORFAFFMDIQRR-UHFFFAOYSA-P
hexafluorosilicon(2-);hydron
21863527
19 to 20.6% (total fluorine)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only