missing translation for 'onlineSavingsMsg'
Learn More

Fluorotriphenylsilane 97.0+%, TCI America™

Catalog No. p-7129686
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04395G 5 g
F043925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04395G Supplier TCI America Supplier No. F04395G

Chemical Identifiers

CAS 379-50-0
Molecular Formula C18H15FSi
Molecular Weight (g/mol) 278.401
MDL Number MFCD00017899
InChI Key UBGXLEFOIVWVRP-UHFFFAOYSA-N
Synonym Triphenylfluorosilane, Triphenylsilyl Fluoride
PubChem CID 67837
IUPAC Name fluoro(triphenyl)silane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)F

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 210°C
Quantity 5 g
UN Number 3261
Formula Weight 278.40
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Fluorotriphenylsilane