Learn More
Invitrogen™ Fluoromount-G™ Mounting Medium
Catalog No. 5018788
Change view
Click to view available options
Quantity:
25 mL
Catalog No. Quantity
5018788 25 mL
1 options
Catalog No. 5018788 Supplier Invitrogen™ Supplier No. 00495802
  • ONLINE EXCLUSIVE PRICE
    $144.00
    $118.08 / Each
    Save $25.92
    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed for use in mounting slides following immunofluorescent staining. Water-soluble Fluoromount-G™ is used to mount slides in which the final step of staining is aqueous. It forms a semi-permanent seal for prolonged storage of slides at 2° to 8°C.
  • Fluoromount-G™ does not fluoresce and may reduce the amount of fluorochrome quenching during fluorescence microscopy
  • Clear liquid medium
  • Supplied as a 1x stock solution containing sodium azide
Specifications
Quantity 25 mL
Product Type Mounting Medium
Product Line Fluoromount-G
Reagent Type Mounting Solution
For Research Use Only. Not for use in diagnostic procedures.