Learn More
Invitrogen™ Fluoromount-G™ Mounting Medium, with DAPI
Catalog No. 501128966
Change view
Click to view available options
Quantity:
20 mL
Catalog No. Quantity
501128966 20 mL
1 options
Catalog No. 501128966 Supplier Invitrogen™ Supplier No. 00495952
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use in mounting slides following immunofluorescent staining as well as staining double stranded DNA. Water-soluble Fluoromount-G™ is used to mount slides in which the final step of staining is aqueous
  • Fluoromount-G™, with DAPI has a maximal fluorescent emission at 455nm and may reduce the amount of fluorochrome quenching during fluorescence microscopy
  • Clear liquid medium containing a fluorescent nuclear stain
  • Supplied as a 1x stock solution containing sodium azide
Specifications
Quantity 20 mL
Product Type Mounting Medium
Product Line Fluoromount-G
Reagent Type Mounting Solution
For Research Use Only. Not for use in diagnostic procedures.