missing translation for 'onlineSavingsMsg'
Learn More

Fluorometholone, 97%, Thermo Scientific Chemicals

Catalog No. AC460200010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460200010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460200010 Supplier Thermo Scientific Chemicals Supplier No. 460200010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorometholone, CAS # 426-13-1, is a glucocorticoid with anti-inflammatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Fluorometholone is a synthetic glucocorticoid
  • It has anti-inflammatory properties

Applications

  • Fluorometholone interacts with cytoplasmic glucocorticoid receptors resulting in activation of these receptors, which mediates gene expression

Chemical Identifiers

CAS 426-13-1
Molecular Formula C22H29FO4
Molecular Weight (g/mol) 376.46
InChI Key FAOZLTXFLGPHNG-KNAQIMQKSA-N
Synonym fluorometholone, oxylone, flumetholon, fluormetholone, fml liquifilm, fluor-op, cortilet, delmeson, trilcin, fml forte
PubChem CID 9878
ChEBI CHEBI:31625
IUPAC Name (6S,8S,9R,10S,11S,13S,14S,17R)-17-acetyl-9-fluoro-11,17-dihydroxy-6,10,13-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES CC1CC2C3CCC(C3(CC(C2(C4(C1=CC(=O)C=C4)C)F)O)C)(C(=O)C)O

Specifications

Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information (2.5% in pyridine) clear colorless to light yellow
Formula Weight 376.46
Percent Purity 97%
Chemical Name or Material Fluorometholone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
EINECSNumber 207-041-5

RUO – Research Use Only