missing translation for 'onlineSavingsMsg'
Learn More

Fluorolink∣r E10-H

Use in polymer modifications | CAS: 162492-15-1

$117.40 - $370.71

Chemical Identifiers

CAS 162492-15-1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6637414
View Documents
Thermo Scientific Chemicals
H6637414
25 g
Each for $117.40
Only null left
 
AAH6637422
View Documents
Thermo Scientific Chemicals
H6637422
100 g
Each for $370.71
Only null left
 
Description

Description

Use in polymer modifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use in polymer modifications.

Solubility
Soluble in fluorinated solvents, alcohol, and ketones. Insoluble in water.

Notes
Incompatible with heat and oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications

Chemical Identifiers

162492-15-1

Specifications

162492-15-1
1.73 g/mL
1.317
Soluble in fluorinated solvents,alcohol,and ketones; Insoluble in water.
Fluorolink™ E10-H
Colorless to Yellow
Odorless
25 g
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 500-749-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only