missing translation for 'onlineSavingsMsg'
Learn More

Fluorocyclohexane 98.0+%, TCI America™

Catalog No. F05875G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05875G Supplier TCI America Supplier No. F05875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 372-46-3
Melting Point 13°C
Color Yellow
Boiling Point 104°C
Molecular Formula C6H11F
Quantity 5 g
MDL Number MFCD00021285
UN Number 1993
Synonym cyclohexane, fluoro, cyclohexyl fluoride, cyclohexylfluoride, fluoro-cyclohexane, acmc-1cpcv
InChI Key GOBGVVAHHOUMDK-UHFFFAOYSA-N
SMILES C1CCC(CC1)F
IUPAC Name fluorocyclohexane
Molecular Weight (g/mol) 102.152
PubChem CID 78988
Formula Weight 102.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Fluorocyclohexane
Show More Show Less