missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ FluoroBrite™ DMEM
GREENER_CHOICE

Catalog No. p-6543034
Encompass
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1896701 500 mL
A1896702 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1896701 Supplier Gibco™ Supplier No. A1896701
Only null left

Gibco™ FluoroBrite™ DMEM features a background fluorescence that is comparable to PBS and 90% lower than that emitted by standard phenol red–free DMEM.

Gibco™ FluoroBrite™ DMEM features a background fluorescence that is comparable to PBS and 90% lower than that emitted by standard phenol red–free DMEM. Formulated to include the required nutrients for routine cell culture when supplemented with 10% fetal bovine serum and 4 mM L-glutamine or GlutaMAX™ Supplement, FluoroBrite™ DMEM is designed to enhance the signal-to-noise ratio of fluorophores, enabling researchers to visualize even the weakest fluorescent events in an environment that promotes optimum cell health. Additional features include:

  • Enhancement of fluorescence signal during live-cell imaging
  • DMEM-based to help preserve cell health

Live-cell fluorescence microscopy is an essential technique for the visualization of fundamentally important and physiologically relevant biological events. A key challenge with this technique is the ability to image weak fluorophores without causing cell damage, photobleaching, or undesirable changes to cell health. FluoroBrite™ DMEM helps address these issues.

Specifications

Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type DMEM (Dulbecco's Modified Eagle Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives High Glucose
Without Additives No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Quantity 500 mL
Content And Storage Store in refrigerator (2–8°C). Protect from light.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.