missing translation for 'onlineSavingsMsg'
Learn More

Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate, 97%

Catalog No. AC440360050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440360050 5 g Glass bottle
AC440360250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC440360050 Supplier Thermo Scientific Chemicals Supplier No. 440360050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 164298-25-3 | C9H16FN2·F6P | 316.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 164298-25-3
Molecular Formula C9H16FN2·F6P
Molecular Weight (g/mol) 316.2
InChI Key MNJUGQKOHJQOCK-UHFFFAOYSA-N
Synonym btffh, bis tetramethylene fluoroformamidinium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, c9h16fn2.f6p, fluoro-dipyrrolidinocarbenium hexafluorophosphate, 1-fluoro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate t, 1e-1-fluoro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, fluoro-n,n,n inverted exclamation marka,n inverted exclamation marka-bis tetramethylene formamidinium hexafluorophosphate
PubChem CID 10935980
IUPAC Name 1-[fluoro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
SMILES C1CCN(C1)C(=[N+]2CCCC2)F.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 152.0°C to 154.0°C
Color White
Elemental Analysis 32.82 to 35.55% C, 8.51 to 9.21% N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 316.2
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Fluoro-N,N,N′,N′-bis(tetramethylene)formamidinium hexafluorophosphate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only