missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluoro-Max Dyed Green Aqueous Fluorescent Particles

Improve sensitivity and detectability for analytical methods with green aqueous fluorescent particles.

$892.84 - $4294.88

Specifications

Additive Trace amounts of surfactant to inhibit agglomeration and promote stability
Concentration 1% solids
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. Product comes with stir bar for continual mixing duriong sampling.
Contents Dyed polystyrene microspheres in water
Color Green
View More Specs

Products 38
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity  
09980440
View Documents
Thermo Scientific™
G0100
1.0 μm 10 mL
Each for $1,054.53
Only null left
 
09980441
View Documents
Thermo Scientific™
G0100B
1.0 μm 60 mL
Each for $4,294.88
Only null left
 
09980442
View Documents
Thermo Scientific™
G0200
2.0 μm 10 mL
Each for $950.91
Only null left
 
09980443
View Documents
Thermo Scientific™
G0200B
2.0 μm 60 mL
Each for $4,294.88
Only null left
 
09980444
View Documents
Thermo Scientific™
G0220
2.2 μm 10 mL
Each for $950.91
Only null left
 
09980445
View Documents
Thermo Scientific™
G0220B
2.2 μm 60 mL
Each for $4,294.88
Only null left
 
09980446
View Documents
Thermo Scientific™
G0300
3.0 μm 10 mL
Each for $1,054.53
Only null left
 
09980447
View Documents
Thermo Scientific™
G0300B
3.0 μm 60 mL
Each for $4,294.88
Only null left
 
09980448
View Documents
Thermo Scientific™
G0500
5.0 μm 10 mL
Each for $950.91
Only null left
 
09980449
View Documents
Thermo Scientific™
G0500B
5.0 μm 60 mL
Each for $4,294.88
Only null left
 
09-980-416
View Documents
Thermo Scientific™
G100
0.10 μm 15 mL
Each for $892.84
Only null left
 
09980451
View Documents
Thermo Scientific™
G1000B
10.0 μm 60 mL
Each for $4,294.88
Only null left
 
09980417
View Documents
Thermo Scientific™
G100B
0.10 μm 90 mL
Each for $4,294.88
Only null left
 
09-980-420
View Documents
Thermo Scientific™
G200
0.20 μm 15 mL
Each for $892.84
Only null left
 
09980421
View Documents
Thermo Scientific™
G200B
0.20 μm 90 mL
Each for $4,294.88
Only null left
 
09980406
View Documents
Thermo Scientific™
G25
0.03 μm 15 mL
Each for $950.91
Only null left
 
09980422
View Documents
Thermo Scientific™
G250
0.25 μm 15 mL
Each for $950.91
Only null left
 
09980423
View Documents
Thermo Scientific™
G250B
0.25 μm 90 mL
Each for $4,294.88
Only null left
 
09980407
View Documents
Thermo Scientific™
G25B
0.03 μm 90 mL
Each for $4,294.88
Only null left
 
09980424
View Documents
Thermo Scientific™
G300
0.30 μm 15 mL
Each for $950.91
Only null left
 
09980425
View Documents
Thermo Scientific™
G300B
0.30 μm 90 mL
Each for $4,294.88
Only null left
 
09-980-408
View Documents
Thermo Scientific™
G40
0.04 μm 15 mL
Each for $892.84
Only null left
 
09980427
View Documents
Thermo Scientific™
G400B
0.40 μm 90 mL
Each for $4,294.88
Only null left
 
09980409
View Documents
Thermo Scientific™
G40B
0.04 μm 90 mL
Each for $4,294.88
Only null left
 
09980410
View Documents
Thermo Scientific™
G50
0.05 μm 15 mL
Each for $950.91
Only null left
 
09980430
View Documents
Thermo Scientific™
G500
0.50 μm 15 mL
Each for $1,054.53
Only null left
 
09980431
View Documents
Thermo Scientific™
G500B
0.50 μm 90 mL
Each for $4,294.88
Only null left
 
09980411
View Documents
Thermo Scientific™
G50B
0.05 μm 90 mL
Each for $4,294.88
Only null left
 
09980433
View Documents
Thermo Scientific™
G600B
0.60 μm 90 mL
Each for $4,294.88
Only null left
 
09980435
View Documents
Thermo Scientific™
G700B
0.70 μm 90 mL
Each for $4,294.88
Only null left
 
09-980-412
View Documents
Thermo Scientific™
DUKG75
0.08 μm 15 mL
Each for $892.84
Only null left
 
09980413
View Documents
Thermo Scientific™
G75B
0.08 μm 90 mL
Each for $4,294.88
Only null left
 
09980414
View Documents
Thermo Scientific™
G85
0.09 μm 15 mL
Each for $950.91
Only null left
 
09980415
View Documents
Thermo Scientific™
G85B
0.09 μm 90 mL
Each for $4,294.88
Only null left
 
09980438
View Documents
Thermo Scientific™
G900
0.90 μm 15 mL
Each for $950.91
Only null left
 
09980439
View Documents
Thermo Scientific™
G900B
0.90 μm 90 mL
Each for $4,294.88
Only null left
 
09980437
View Documents
Thermo Scientific™
G830B
0.83 μm 90 mL
Each for $4,293.38
Only null left
 
09-980-450
View Documents
Thermo Scientific™
G1000
10.0 μm 10 mL
Each for $940.19
Only null left
 
Description

Description

Improve sensitivity and detectability for analytical methods with Thermo Scientific™ Fluoro-Max Green Aqueous Fluorescent Particles, microsphere suspensions that are internally dyed and feature bright, high-contrast colors.

Fluoro-Max fluorescent particles emit bright and distinct colors when illuminated by the light of shorter wavelengths than the emission wavelength, improving their contrast and visibility relative to background materials.These internally dyed polymer particles utilize the exclusive Firefli™ process to incorporate the dye throughout the polymer matrix to produce bright fluorescent colors, minimize photo bleaching and prevent dye leaching into aqueous media.

Features:
  • Made of polystyrene (PS), which has a density of 1.05g/cm3 and a refractive index of 1.59 @ 589nm (25°C)
  • Aqueous suspensions are packaged as 1% solids
  • Can be detected with an epifluorescence microscope, confocal microscope, fluorometer, fluorescence spectrophotometer or fluorescence activated cell sorter
  • Can also be detected using mineral light or black light (UV)
Note:

Spectral information is approximate and for reference only. Spectral properties of the dyes are dependent on their concentration and physical environment.The exact excitation and emission maxima may vary depending on the size and composition of the particles.

Recommended for:

  • Filter testing
  • Flow tracing
  • Biological research
Specifications

Specifications

Trace amounts of surfactant to inhibit agglomeration and promote stability
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. Product comes with stir bar for continual mixing duriong sampling.
Green
Fluorescent Particle
Package Insert Sheet
1.59 at 589nm (25°C)
1% solids
Dyed polystyrene microspheres in water
Polystyrene
Firefli™ Fluorescent Green (468/508nm)
1.05g/cm³
Product stable for a minimum of 24 months
Videos
SDS
Documents

Documents