missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Fluorescent Shop Envelopes

Color-code work in process, machine procedures and drawings. Brady™ Fluorescent Shop Envelopes are the perfect solution for organizing work flows.

$197.52

Specifications

Material Plastic
Mounting Type Metal Grommet
For Use With (Application) Color-coding work in process, machine procedures and drawings
Quantity 25/Pk.
Dimensions (L x W) 9 x 6 in.
View More Specs

Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19113411
View Documents Promotion Details
Brady™
56944
Fluorescent Orange Trim on Clear
Pack of 25 Each for $197.52
Only null left
 
Description

Description

  • Fluorescent trimmed shop envelopes
  • Ideal for color-coding work in process, machine procedures and drawings
  • For indoor and outdoor use
Specifications

Specifications

Plastic
Color-coding work in process, machine procedures and drawings
9 x 6 in.
6 in.
Metal Grommet
25/Pk.
9 in.
Videos
SDS
Documents

Documents